Learn More
IKA™ Dry Block Heater 4
Sfoglia Tutto IKA Prodotti
Click to view available options
Tensione:
220/240 V, 115/100 V
Dimensione della confezione:
Pezzo
Descrizione
- Digital block heater for four aluminum blocks
- Included PT-1000 temperature sensor allows temperature control directly in the sample vessel
- Block heater is ideal for melting and boiling point determination, enzyme reactions, incubation and activation of cultures, immunoassays, DNA denaturation, culture media tests, coagulation tests, blood-urea-nitrogen determinations and in situ hybridizations
- Stepless adjustable temperature up to 120°C
- Timer: Countdown, adjustable from 1 min. to 99h 59 min.
- Counter: Displays heating time
- Highly versatile with a large number of blocks
- Error code display
- Fixed safety circuit
- Acoustic signal at end of test
Specifica
Specifica
| Temperature Range (Metric) | Ambient to 120°C |
| Tipo | Bagno secco/blocco |
| N. di blocchi | 4 |
| Larghezza (sistema metrico) | 151 mm |
| Altezza (sistema metrico) | 73 mm |
| Requisiti elettrici | 220/240 V, 115/100 V, 50/60 Hz |
| Tensione | 220/240 V, 115/100 V |
| Wattage | 412 w |
| Frequenza | 50/60 Hz |