missing translation for 'onlineSavingsMsg'
Learn More

Grant Instruments™ Dry-Block Heater/Cooler

Codice prodotto. p-7241268
Click to view available options
Dimensioni provetta:
1.5 mL
2 mL
Dimensione della confezione:
Pezzo
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 17795019

Marca: Grant Instruments™ PCH2 12V

Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

The Grant Instruments PCH-1/PCH-2/PCH-3 personal benchtop cooler/heaters are ideal for laboratory applications requiring cooling and heating in microtubes within the temperature setting range of – 10°C to +100°C.

The PCH cooler/heaters enable a stable temperature to be maintained during room temperature fluctuations.
  • The ingenious block construction, combined with a powerful Peltier module, produces very rapid cooling and heating for swift change of application and increased flexibility. In addition, the Peltier effect produces efficient cooling and heating for small thermal loads without the use of a refrigeration system.
  • The dry temperature control system maintains a clean working environment, reducing the chance of contamination or sample dilution. A Peltier system, without the vibration of refrigeration or the turbulence of a waterbath, is also ideal for all incubations where stable conditions are critical.
  • The PCH is a very effective tool for DNA sample preparation for denaturing electrophoresis. The exceptionally rapid heat up enables the denaturation temperature of 95°C to be reached swiftly, the timer will signify denaturation time-up, and the samples can be rapidly cooled again ready to load the gels.
  • Cooling/heating setting range from – 10ºC to 100°C, with very rapid cool down and heat-up times
  • Stability ± 0.1°C, uniformity ±0.25°C at 37°C
  • Choice of two models; capacity for up to 32 microtubes in a combination of two sizes: 12 x 1.5 mL plus 20 x 0.5 mL (PCH-1) or up to 20 microtubes of one size: 20 x 1.5 mL tubes (PCH-2), up to a total of 20 x 2 mL microtubes (PCH-3)
  • Convenient integral reaction timer with audible alarm
  • Dry temperature control system maintains a clean and vibration-free working environment
  • A 2-line LCD display clearly indicates both set and actual values for temperature and time. Works in combination with simple push buttons for easy set-up.
  • Flexible tube sizes and rapid heat-up/cool-down times enables swift change of application

Applications:

Life science – storing frozen restriction enzymes, nick translations, ligation reactions, restriction digests, protein solubilisation for PAGE, warm incubation of microcentrifuge tubes for hybridisation, enzyme reactions and deactivations.

Clinical – cooling blood samples prior to coagulation testing.

TRUSTED_SUSTAINABILITY

Specifica

Colore White
Descrizione Dry-block heater/cooler for 20 x 1.5 mL microtubes -10°C to +100°C (ambient -30°C minimum)
Capacità 20 Microtubes
Tipo di spina US, EU, UK, AUS Plug
Temperature Range (Metric) -10°C to +100°C (Cooling/Heating Setting Range)
Tipo Dry-Block Heater/Cooler
Display 2 line x 16 character LCD
Modello PCH-2 12 V
N. di blocchi 1
Stabilità termica Stability ±0.1°C, uniformity ±0.25°C at 37°C
Temporizzatore 1 min. to 96 hr., Non-stop
Dimensioni provetta 1.5 mL
Warranty 2 years
Dimensioni (L x P x A) 32 x 30 x 25 cm
Peso di spedizione (sistema metrico) 4 kg
Peso (sistema metrico) 3.2 kg
Tensione 12 V
Wattage 55 W
Amperaggio 4.4 A
Vedi altri risultati Mostra meno risultati
Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato