Learn More
BUCHI Lyovapor™ Drying Chamber
Sfoglia Tutto Buchi Prodotti
Click to view available options
Tipo:
Benchtop Freeze Dryer
Heated Product Shelf Chamber
N. di ripiani:
0
4
6
Stile:
12-position Manifold Top
24-position Manifold Top
Standard Top
Stoppering Vials Top
Include:
4 Sample Temperature Sensors
6 Sample Temperature Sensors
Chamber Only
Drying Chamber Only
Pressure Difference Testing
Dimensione della confezione:
Pezzo
Descrizione
- Manifolds: adapters for traditional freeze drying beakers or round bottom flasks
- Shelves: for vials, plates, and irregular shaped items
- Heated Shelves: advanced control of drying process and applications
- Stoppering Chambers: seal vials without exposure to ambient moisture
Specifica
Specifica
| Tipo | Benchtop Freeze Dryer |
| N. di ripiani | 4 |
| Temperature Range (Metric) | Ambient to -105°C |
| Materiale | Acrylic |
| Stile | Standard Top |
| Include | Drying Chamber Only |
| Tipo di spina | North America |
| Tensione | 220/240 V |