missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BUCHI Drying Rack Manifold 12
Shop All Buchi Products
Click to view available options
Tipo di spina:
North America
Unit Size:
Pezzo
Description
- Manifolds: adapters for traditional freeze drying beakers or round bottom flasks
- Shelves: for vials, plates, and irregular shaped items
- Heated Shelves: advanced control of drying process and applications
- Stoppering Chambers: seal vials without exposure to ambient moisture
Specifications
Specifications
| Tipo | Benchtop Freeze Dryer |
| N. di ripiani | 0 |
| Temperature Range (Metric) | Ambient to -105°C |
| Materiale | Stainless Steel |
| Tipo di spina | North America |
| Tensione | 220/240 V |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction