Learn More
dsDNA Antibody (rDSD/8266), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP3-24019PE
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
dsDNA Monoclonal antibody specifically detects dsDNA in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifica
| dsDNA | |
| Monoclonal | |
| PE | |
| Double stranded DNA | |
| Nuclei of Burkitt's cells | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Immunohistochemistry (Paraffin) | |
| rDSD/8266 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Human | |
| Purified |