Learn More
Fisherbrand™ Dual-Purpose Pipets
Codice prodotto. p-164364 Sfoglia Tutto Fisherbrand Prodotti
Click to view available options
Volume (sistema metrico):
1 mL
2 mL
5 mL
Accuracy:
±0.006 mL
±0.02 mL
Dimensione della confezione:
pezzi 12
Codice prodotto. 16650093
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Dual-purpose pipets
  • Borosilicate glass, permanent amber markings, Tuf-Temp™-treated to reduce chipping
  • Accuracy printed on each pipet
  • Capacity marked below mouthpiece
Order Info
Available as PersonaPet™ personalized pipets
Compliance
Calibrated To Deliver/Blow Out (upper graduation) and To Contain (lower graduation).
Specifica
Accuracy ±0.006 mL
Materiale Borosilicate Glass
Volume (sistema imperiale) 0.03 oz.
Volume (sistema metrico) 1 mL
Tipo Dual Purpose Pipet