Learn More
VEE GEE Dual Scale Hydrometer
Click to view available options
Intervallo Baumé:
0° to 26° (Heavy)
0° to 41° (Heavy)
0° to 45° (Heavy)
0° to 72° (Heavy)
10° to 100° (Light)
24° to 43 (Heavy)
25° to 42° (Heavy)
42° to 55° (Heavy)
55° to 65° (Heavy)
64° to 72° (Heavy)
70° to 10° (Light)
Intervallo di gravità specifica:
0.600 to 1.000
0.70 to 1.00
1.000 to 1.220
1.000 to 1.225
1.000 to 1.400
1.000 to 1.450
1.000 to 2.000
1.200 to 1.420
1.200 to 1.425
1.400 to 1.620
1.600 to 1.820
1.800 to 2.020
Dimensione della confezione:
Pezzo
Descrizione
- The Dual Scale Specific Gravity/Baume Hydrometer includes two scales.
- One scale is designed for liquids heavier than water.
- The other scale is specifically for liquids lighter than water.
Specifica
Specifica
| Tipo | Specific Gravity and Baume Hydrometer |
| Incrementi | 0.005/1° |
| Lunghezza (sistema metrico) | 30.5 cm |
| Intervallo Baumé | 10° to 100° (Light) |
| Intervallo di gravità specifica | 0.600 to 1.000 |
| Temperature of Standardization | 15.6°C (60°F) |