Learn More
DWK Life Sciences DURAN™ Adapter, standard ground joint
Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Tipo:
Expansion Joint
Expansion Joint, Non-DIN Size
Reduction Joint
Reduction Joint, Non-DIN Size
Joint Size:
Socket NS 14/23, Cone NS 19/26
Socket NS 14/23, Cone NS 24/29
Socket NS 14/23, Cone NS 29/32
Socket NS 19/26, Cone NS 14/23
Socket NS 19/26, Cone NS 24/29
Socket NS 19/26, Cone NS 29/32
Socket NS 24/29, Cone NS 14/23
Socket NS 24/29, Cone NS 19/26
Socket NS 24/29, Cone NS 29/32
Socket NS 29/32, Cone NS 14/23
Socket NS 29/32, Cone NS 19/26
Socket NS 29/32, Cone NS 24/29
Dimensione della confezione:
Pezzo
Descrizione
Transition adapters or reducers are glass components with two standard ground joints of different sizes. With the help of these adapters, glass components with different standard ground joints can be connected to each other.
- Tight closure due to standard ground joint
Specifica
Specifica
| Quantità | 1 Ea. |
| Descrizione articolo | Connection of two glass components |
| Tipo | Expansion Joint, Non-DIN Size |
| Joint Size | Socket NS 14/23, Cone NS 24/29 |