Learn More
Lenz Laborglasinstrumente™ Palloni a tre colli in vetro borosilicato DURAN™ con colli laterali inclinati
Especially made for distillation, reactions and to make solutions or hold, contain and collect chemicals. Lenz Laborglasinstrumente™ Three-Neck Round Bottom Flasks, Side Necks Angled are manufactured from durable DURAN™ borosilicate glass.
Marca: Lenz Laborglasinstrumente™ 03314158
Dettagli aggiuntivi : Peso : 0.23400kg
Descrizione
- Acc. to DIN 12394
- Two side necks at 20° angles
- With conical ground joints, featuring high taper accuracy and excellent tightness
- With inscription label
- Corrosion-free and resistant to thermal shock
Specifica
500 mL | |
Vetro borosilicato DURAN™, Duran Glass | |
14/23 NS collo lato, 29/32 NS collo centro | |
Rotondo |