Learn More
Lenz Laborglasinstrumente™ Palloni a tre colli in vetro borosilicato DURAN™ con colli laterali inclinati
Codice prodotto. 10382701
Click to view available options
Capacità (sistema metrico):
25 mL
100 mL
250 mL
500 mL
1000 mL
5 L
Giunto a bussola:
14/23 NS Side Neck, 24/29 NS Center Neck
14/23 NS collo lato, 14/23 NS collo centro
14/23 NS collo lato, 29/32 NS collo centro
19/26 NS Side Neck, 24/29 NS Center Neck
19/26 NS Side Neck, 29/32 NS Center Neck
29/32 NS Side Neck, 29/32 NS Center Neck
Dimensione della confezione:
Pezzo
Codice prodotto. 10382701
Marca: Lenz Laborglasinstrumente™ 03314249
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Especially made for distillation, reactions and to make solutions or hold, contain and collect chemicals. Lenz Laborglasinstrumente™ Three-Neck Round Bottom Flasks, Side Necks Angled are manufactured from durable DURAN™ borosilicate glass.
  • Acc. to DIN 12394
  • Two side necks at 20° angles
  • With conical ground joints, featuring high taper accuracy and excellent tightness
  • With inscription label
  • Corrosion-free and resistant to thermal shock
Specifica
Capacità (sistema metrico) 250 mL
Materiale Borosilicate Glass, Duran Glass
Giunto a bussola 19/26 NS Side Neck, 29/32 NS Center Neck
Forma del fondo Round