Learn More
Lenz Laborglasinstrumente™ Palloni a tre colli in vetro borosilicato DURAN™ con colli laterali paralleli
Codice prodotto. p-8006016
Click to view available options
Capacità (sistema metrico):
100 ml
250 mL
500 ml
1000 mL
2000 mL
4 L
6 L
10 L
Giunto a bussola:
14/23 NS Side Neck, 29/32 NS Center Neck
19/26 NS Side Neck, 29/32 NS Center Neck
29/32 NS Side Neck, 29/32 NS Center Neck
29/32 NS Side Neck, 45/40 NS Center Neck
29/32 NS collo lato, 29/32 NS collo centro
Dimensione della confezione:
Pezzo
Codice prodotto. 10720992
Marca: Lenz Laborglasinstrumente™ 03305488
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Especially made for distillation, reactions and to make solutions or hold, contain and collect chemicals. Lenz Laborglasinstrumente™ Three-Neck Round Bottom Flasks, Side Necks Parallel are manufactured from durable DURAN™ borosilicate glass.
  • Acc. to DIN 12392
  • Two parallel side necks at 90° angle
  • With conical ground joints, featuring high taper accuracy and excellent tightness
  • With inscription label
  • Corrosion-free and resistant to thermal shock
Specifica
Capacità (sistema metrico) 10 L
Materiale Vetro borosilicato Duran™, Duran Glass
Giunto a bussola 29/32 NS Side Neck, 45/40 NS Center Neck
Forma del fondo Rotondo