Learn More
DWK Life Sciences DURAN™ Centrifuge Tube, with round bottom
Codice prodotto. 11786908 Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Capacità (sistema metrico):
6 mL
12 mL
25 mL
50 mL
80 mL
250 mL
Diametro (sistema metrico):
12 mm
16 mm
24 mm
34 mm
40 mm
44 mm
56 mm
Dimensione della confezione:
pezzi 10
pezzi 50
Codice prodotto. 11786908
Marca: DWK Life Sciences 216011707
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Centrifuge tubes are used for centrifuging. Here a separation of the mixture of substances takes place due to the density of the individual components.
One of the most important basic process engineering methods is the separation of mixtures of substances. This working method includes centrifugation, which exploits the concept of mass inertia. A mixture of substances to be separated is filled into centrifuge tubes, placed in a centrifuge and rotated. Due to the high rotation speed, heavier particles are pushed outwards faster and further away than the lighter ones. This results in separation based on the mass or density of the individual material components.
  • Very resistant to mechanical loading
  • Round bottom allows sedimentation/separation of solids on a larger surface
TRUSTED_SUSTAINABILITY
Specifica
Capacità (sistema metrico) 50 mL
Materiale Borosilicate Glass (3.3)
Da utilizzare con Centrifugation; Separation of different particles due to different mass inertia (density); Separation of materials by utilization of centrifugal force
Quantità 10/Pk.
Autoclavabile Autoclavable
Diametro (sistema metrico) 34 mm