Learn More
DWK Life Sciences DURAN™ Centrifuge Tube, with round bottom
Sfoglia Tutto DWK Life Sciences ProdottiDescrizione
One of the most important basic process engineering methods is the separation of mixtures of substances. This working method includes centrifugation, which exploits the concept of mass inertia. A mixture of substances to be separated is filled into centrifuge tubes, placed in a centrifuge and rotated. Due to the high rotation speed, heavier particles are pushed outwards faster and further away than the lighter ones. This results in separation based on the mass or density of the individual material components.
- Very resistant to mechanical loading
- Round bottom allows sedimentation/separation of solids on a larger surface
Specifica
Specifica
| Capacità (sistema metrico) | 80 mL |
| Materiale | Borosilicate Glass (3.3) |
| Da utilizzare con | Centrifugation; Separation of different particles due to different mass inertia (density); Separation of materials by utilization of centrifugal force |
| Quantità | 10/Pk. |
| Autoclavabile | Autoclavable |
| Diametro (sistema metrico) | 40 mm |
| Product Type | Round Bottom Centrifuge Tube |