Learn More
DWK Life Sciences DURAN™ Expansion Adapter, socket/cone
Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Joint Size:
Socket NS 14/23, Cone NS 19/26
Socket NS 14/23, Cone NS 24/29
Socket NS 14/23, Cone NS 29/32
Socket NS 19/26, Cone NS 24/29
Socket NS 19/26, Cone NS 29/32
Socket NS 24/29, Cone NS 29/32
Socket NS 24/29, Cone NS 45/40
Socket NS 29/32, Cone NS 34/35
Socket NS 29/32, Cone NS 45/40
Socket NS 29/32, Cone NS 50/42
Socket NS 29/32, Cone NS 60/46
Socket NS 29/32, Cone NS 71/51
Dimensione della confezione:
Pezzo
pezzi 10
Descrizione
Adapters are used to connect two components with different NS sizes. DURAN™ expansion pieces allow the connection of a smaller sleeve to a larger core.
- Expansion pieces allow the connection of a smaller sleeve to a larger core and thus the use of differently sized ground joint components.
Specifica
Specifica
| Quantità | 10/Pk. |
| Descrizione articolo | Connection of two components with different standard ground joint sizes |
| Joint Size | Socket NS 24/29, Cone NS 29/32 |