Learn More
DWK Life Sciences DURAN™ Funnel, with long stem, Bunsen Funnel
Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Diametro (sistema metrico) parte superiore:
55 mm
70 mm
80 mm
Dimensione della confezione:
pezzi 10
Descrizione
Transferring liquids and solids is a common working method. In this activity the funnel is used, which due to its shape can absorb a relatively large amount of liquid or solid and transfer it through a constriction into a vessel with a small opening. Funnels are available in different sizes and designs, selected according to their application.
- With long stem
- Bunsen funnel
- Fired-on, highly durable white print
Specifica
Specifica
| Diametro (sistema metrico) parte superiore | 80 mm |
| Materiale | Borosilicate Glass (3.3) |
| Autoclavabile | Autoclavable |
| Da utilizzare con (applicazione) | Decanting and filtration of liquids of different densities; The spillage of substances is avoided; Filtering |
| Quantità | 10/Pk. |
| Product Type | Non-DIN/ISO size |