missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ GLS 80™ Laboratory Bottle, Wide Mouth, Clear, GLS 80™ Thread, Graduated, Bottle Only
Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Capacità (sistema metrico):
10,000 mL
1000 mL
20,000 mL
2000 mL
250 mL
3500 mL
500 mL
5000 mL
Dimensione della confezione:
Pezzo
pezzi 10
Descrizione
™™ A combination of high-performance materials and robust ISO 4796-1 design provides a long working life. The bottles feature an easy-to-read graduated volume scale, and marking field in permanent white enamel. Permanent Retrace Code on each bottle for manufacturing lot traceability. ™™° °
- Easy access to the bottle contents due to the wide diameter of the bottle neck
- Special thread design for rapid opening and closing in less than a turn
- Optimum filling and pouring and particularly easy cleaning
- Spoons, spatulas and tweezers can be manipulated with ease
- Even large-volume funnels can be inserted into the bottles without any difficulty
- Stability due to the large base area
- Easy-to-read scale with graduation marks to estimate the content
- Fired-on, highly durable white ceramic
- Complete with blue screw cap (PP, integral lip seal) and pouring ring (PP) for drip-free pouring and clean, safe working
Specifica
Specifica
| Capacità (sistema metrico) | 500 mL |
| Da utilizzare con (applicazione) | Autoclaving of media and reagents; Preparation of media and reagents; Sample collection; Culturing vessel; Storage of media and reagents; Transport and shipping; Liquid handling reservoir |
| Include | Senza tappo a vite e anello antigoccia in PP (blu) |
| Materiale | Borosilicate Glass (3.3) |
| Quantità | 10/Pk. |
| Dimensioni filettatura (sistema metrico) | GLS 80 |
| Materiale chiusura | Polypropylene |
| Diametro (sistema metrico) | 101 mm |
| Autoclavabile | Autoclavable |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto