Learn More
DWK Life Sciences DURAN™ Hexagonal Stopper, pointed, diamond-ground
Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Size:
NS 10/19
NS 12/21
NS 14/23
NS 19/26
NS 24/29
NS 29/32
NS 34/35
NS 45/40
NS 7/16
Dimensione della confezione:
Pezzo
pezzi 10
pezzi 100
Descrizione
DURAN™ hexagonal hollow plugs are used for sealing laboratory equipment such as volumetric flasks, mixing cylinders, ground joint flasks, etc. DURAN™ hexagonal hollow plugs are available in 2 standard versions - pointed - flat - Sizes analogous to DURAN™ conical ground joints. Cut quality: DG = Diamond Ground
- Pointed
- Diamond-ground
- With a dripping syringe, for example, a condensate can be collected and the reaction medium recirculated.
- hollow glass stopper
Specifica
Specifica
| Da utilizzare con (applicazione) | Closure of vessels with standard ground joints |
| Quantità | 100/Pk. |
| Size | NS 24/29 |
| Autoclavabile | Autoclavable |
| Materiale | Borosilicate Glass |