Learn More
DWK Life Sciences DURAN™ Hexagonal Stopper, short ground, flat, finished
Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Size:
NS 10/13
NS 12/14
NS 14/15
NS 19/17
NS 24/20
NS 29/22
Dimensione della confezione:
pezzi 100
Descrizione
DURAN™ hexagonal hollow plugs as a short ground version suitable for DURAN™ short ground sleeves. Cut quality: Ground.
- Short grinding
- Flat
- Ground
- Hollow glass stopper
Specifica
Specifica
| Da utilizzare con (applicazione) | Closure of vessels with standard ground joints; Used for closing short ground sockets |
| Quantità | 100/Pk. |
| Size | NS 14/15 |
| Autoclavabile | Autoclavable |
| Materiale | Borosilicate Glass |