Learn More
DWK Life Sciences DURAN™ KPG™ Stirrer Bearing, interchangeable
Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Joint Size:
HA 10
HB 10
HB 16
Tipo:
Ground Joint
Polished
Dimensione della confezione:
pezzi 10
pezzi 50
Descrizione
DURAN™ KPG™ bearing sleeves are used as components for agitator guides/agitator coolers. They are available as ground or polished versions in the following designs: HA: without grease cup HB: with grease cup
- Available as ground or polished version in the following designs: HA: without grease cup HB: with grease cup
- Small gap dimensions allow a completely smooth running and working under vacuum.
- The bearing sleeve and agitator shaft are precisely matched to each other by precision grinding.
Specifica
Specifica
| Quantità | 10/Pk. |
| Descrizione articolo | As component for stirrer guides / stirrer coolers |
| Tipo | Polished |
| Joint Size | HA 10 |