Learn More
DWK Life Sciences DURAN™ KPG™ Stirrer Shaft, interchangeable
Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Tipo:
Ground Joint
Polished
Dimensione della confezione:
pezzi 50
Descrizione
Solid DURAN™ KPG™ stirrer shafts are used for further processing into a wide variety of stirrers. They are available in the qualities ground and polished. The bearing sleeve and agitator shaft are precisely matched to each other by precision grinding.
- Solid
- Available in polished and ground version
- The bearing sleeve and agitator shaft are precisely matched to each other by precision grinding
Specifica
Specifica
| Quantità | 50/Pk. |
| Descrizione articolo | Stirring of reaction solutions with high viscosities or at high stirring speeds; For further processing to various stirrer shapes and types |
| Tipo | Ground Joint |