Learn More
DWK Life Sciences DURAN™ Long Ground Socket, finished
Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Joint Size:
17 mm
22 mm
28 mm
33 mm
Dimensione della confezione:
pezzi 50
Descrizione
DURAN™ sleeves in the long-grinding variant are frequently used in the high-vacuum range.
- Ground (finer grinding quality, less roughness depth)
- Short grinding for apertures with low space availability and lower tightness requirements
- Tight closure due to standard ground joint
- Glass articles for further processing
Specifica
Specifica
| Quantità | 50/Pk. |
| Descrizione articolo | Use in the high vacuum range; Installation in glass apparatus and devices |
| Joint Size | 17 mm |
| Spessore (sistema metrico) parete | 1.8 mm |