Learn More
DWK Life Sciences DURAN™ Long Ground Socket, finished
Codice prodotto. 15874818 Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Joint Size:
17 mm
22 mm
28 mm
33 mm
Dimensione della confezione:
pezzi 50
Codice prodotto. 15874818
Marca: DWK Life Sciences 286546209
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
For ground joints in the high vacuum range.
DURAN™ sleeves in the long-grinding variant are frequently used in the high-vacuum range.
  • Ground (finer grinding quality, less roughness depth)
  • Short grinding for apertures with low space availability and lower tightness requirements
  • Tight closure due to standard ground joint
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifica
Quantità 50/Pk.
Descrizione articolo Use in the high vacuum range; Installation in glass apparatus and devices
Joint Size 28 mm
Spessore (sistema metrico) parete 2 mm