Learn More
DWK Life Sciences DURAN™ Reduction Adapter, socket/cone
Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Joint Size:
Socket NS 19/26, Cone NS 14/23
Socket NS 24/29, Cone NS 14/23
Socket NS 24/29, Cone NS 19/26
Socket NS 29/32, Cone NS 14/23
Socket NS 29/32, Cone NS 19/26
Socket NS 29/32, Cone NS 24/29
Dimensione della confezione:
Pezzo
pezzi 10
Descrizione
Adapters are used to connect two components with different NS sizes. DURAN™ reducers allow the connection of a larger sleeve to a smaller core.
- Reducers allow the connection of a larger sleeve to a smaller core and thus the use of differently sized ground joint components.
Specifica
Specifica
| Quantità | 10/Pk. |
| Descrizione articolo | Connection of two components with different standard ground joint sizes |
| Joint Size | Socket NS 24/29, Cone NS 14/23 |