Learn More
DWK Life Sciences DURAN™ Screw Thread Tube, GL, heavy wall, supplementary to DIN 12216
Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Nessun giunto rastremato standard:
GL 14
GL 18
GL 25
GL 45
Dimensione della confezione:
Pezzo
pezzi 10
pezzi 100
pezzi 50
Descrizione
Thick-walled DURAN™ glass threaded tubes with GL thread are supplementary to DIN 12216 and are used for attaching to glass apparatus. Available in 5 sizes. The available accessories allow a wide range of applications.
- With standardised GL thread according to DIN 12216
- thick-walled
- Safe and easy sealing and loosening of glass-glass components
- Grease-free working possible
- Glass articles for further processing
Specifica
Specifica
| Quantità | 10/Pk. |
| Descrizione articolo | For attachment to glass apparatuses; Installation in glass apparatus and devices |
| Joint Size | 40 mm |
| Nessun giunto rastremato standard | GL 45 |
| Spessore (sistema metrico) parete | 3.2 mm |