Learn More
DWK Life Sciences DURAN™ Screw Thread Tube, GL, heavy wall, supplementary to DIN 12216
Codice prodotto. 30286581 Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Nessun giunto rastremato standard:
GL 14
GL 18
GL 25
GL 45
Dimensione della confezione:
Pezzo
pezzi 10
pezzi 100
pezzi 50
Codice prodotto. 30286581
Marca: DWK Life Sciences 286256807
For use on a wide variety of apparatus with higher mechanical strength requirements.
Thick-walled DURAN™ glass threaded tubes with GL thread are supplementary to DIN 12216 and are used for attaching to glass apparatus. Available in 5 sizes. The available accessories allow a wide range of applications.
  • With standardised GL thread according to DIN 12216
  • thick-walled
  • Safe and easy sealing and loosening of glass-glass components
  • Grease-free working possible
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifica
Quantità 50/Pk.
Descrizione articolo For attachment to glass apparatuses; Installation in glass apparatus and devices
Joint Size 22 mm
Nessun giunto rastremato standard GL 25
Spessore (sistema metrico) parete 2.5 mm