Learn More
DWK Life Sciences DURAN™ Screw Thread Tube, SVL
Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Nessun giunto rastremato standard:
SVL 15
SVL 22
SVL 30
SVL 42
Dimensione della confezione:
pezzi 10
pezzi 50
Descrizione
The DURAN™ glass thread tube with SVL thread is a finer thread than the GL thread, which is also used for attaching to glass apparatus. The nominal size refers to the core diameter at the thread.
- Glass thread tube with SVL thread is a finer thread compared to GL thread
- The nominal size refers to the core diameter at the thread.
- Safe and easy sealing and loosening of glass-glass components
- Grease-free working possible
- Glass articles for further processing
Specifica
Specifica
| Quantità | 10/Pk. |
| Descrizione articolo | For attachment to glass apparatuses; Installation in glass apparatus and devices |
| Joint Size | 40 mm |
| Nessun giunto rastremato standard | SVL 42 |
| Spessore (sistema metrico) parete | 2 mm |