Learn More
DWK Life Sciences DURAN™ Short Ground Socket, finished
Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Nessun giunto rastremato standard:
NS 14/15
Dimensione della confezione:
pezzi 50
Descrizione
The short-section version of the DURAN™ sleeves is ideal for use where space is limited and tightness requirements are lower.
- According to DIN 12249 Form E
- Ground (finer grinding quality, less roughness depth)
- The finer surface reduces the risk of jamming the cuts.
- Minimized grease requirement
- Very low leakage rate
- Tight closure due to standard ground joint
- Glass articles for further processing
Specifica
Specifica
| Quantità | 50/Pk. |
| Descrizione articolo | Installation in glass apparatus and devices |
| Joint Size | 17 mm |
| Nessun giunto rastremato standard | NS 14/15 |
| Spessore (sistema metrico) parete | 1.8 mm |