Learn More
DWK Life Sciences DURAN™ Single way Stopcock, complete with hollow key with glass handle
Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Dimensione chiusura:
12.5 NS
14.5 NS
18.8 NS
21.5 NS
24 NS
29.2 NS
Dimensione della confezione:
Pezzo
pezzi 25
pezzi 5
pezzi 50
Descrizione
DURAN™ single way stopcocks complete with hollow DURAN™ cock plug are well suited for work with temperature changes and higher tightness requirements.
- Ground shut-off valve sleeve with hollow glass plug with glass handle
- High tightness
- Taper 1:10
- Glass articles for further processing
Specifica
Specifica
| Foro spina | 10 mm |
| Diametro (sistema metrico) | 18 mm |
| Materiale | Borosilicate Glass (3.3) |
| Autoclavabile | Autoclavable |
| Dimensione chiusura | 29.2 NS |
| Larghezza (sistema metrico) | 2.5 mm |
| N. per confezione | 5/Pk. |
| Da utilizzare con (applicazione) | When working with temperature changes and higher tightness requirements; Installation in glass apparatus and devices |