Learn More
DWK Life Sciences DURAN™ Single way Stopcock, ground, without key
Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Foro spina:
1.5 mm
2 mm
2.5 mm
4 mm
6 mm
8 mm
10 mm
Dimensione chiusura:
10 NS
12.5 NS
12.5 NS (1.5 mm width)
14.5 NS
14.5 NS (1.5 mm width)
18.8 NS
21.5 NS
24 NS
29.2 NS
Dimensione della confezione:
pezzi 10
pezzi 25
pezzi 5
pezzi 50
Descrizione
DURAN™ single way stopcocks are designed for use as shut-off valves. Valves are better suited for fine dosing. Ground DURAN™ single way stopcocks can be combined with - DURAN™ tap plug with groove - Threaded tap plug - hollow DURAN™ tap plugs
- Ground shut-off taps without tap plug
- Glass articles for further processing
Specifica
Specifica
| Foro spina | 10 mm |
| Diametro (sistema metrico) | 18 mm |
| Materiale | Borosilicate Glass (3.3) |
| Autoclavabile | Autoclavable |
| Dimensione chiusura | 29.2 NS |
| Larghezza (sistema metrico) | 2.5 mm |
| N. per confezione | 5/Pk. |
| Da utilizzare con (applicazione) | Coarse dosing of liquids; Installation in glass apparatus and devices |