Learn More
DWK Life Sciences DURAN™ Spherical Joint, ball, polished
Codice prodotto. p-7189149 Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Joint Size:
8 mm
12 mm
18 mm
24 mm
28 mm
48 mm
Unit Size:
Pezzo
pezzi 10
Product Code. 15814678
Brand: DWK Life Sciences 286328006
Only null left
Add to basket
Add to basket
For grease-free flexible connections.
The non-sanded version of the DURAN™ spherical ground joint only seals over the sealing ring. Basically 2 versions of the sealing rings are available: - FEP-coated silicone rings - VITON sealing rings The nominal size is composed of the nominal diameter of the spherical ground joint and, if applicable, the clear width of the pipe diameter. This variant also enables flexible connections, these are in the angle range of 10-15°.
  • Flexible connections of up to 15° can be realised depending on the size of the cut (angle ≥ tightness reduced)
  • Grease-free working possible
  • A tight seal is made possible by the use of an additional sealing ring.
  • The nominal size describes the nominal diameter of the spherical ground joint.
  • Not self-supporting
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications
Quantità 10/Pk.
Descrizione articolo For the production of grease-free, flexible connections (alignment in 360° possible); Installation in glass apparatus and devices
Joint Size 28 mm
Spessore (sistema metrico) parete 2.8 mm