Learn More
DWK Life Sciences DURAN™ Spherical Joint, cup, polished
Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Joint Size:
8 mm
12 mm
18 mm
24 mm
28 mm
48 mm
Förpackningsstorlek
Pezzo
pezzi 10
Beskrivelse
The non-sanded version of the DURAN™ spherical ground joint only seals over the sealing ring. Basically 2 versions of the sealing rings are available: - FEP-coated silicone rings - VITON sealing rings The nominal size is composed of the nominal diameter of the spherical ground joint and, if applicable, the clear width of the pipe diameter. This variant also enables flexible connections, these are in the angle range of 10-15°.
- Flexible connections of up to 15° can be realised depending on the size of the cut (angle ≥ tightness reduced)
- Grease-free working possible
- A tight seal is made possible by the use of an additional sealing ring.
- The nominal size describes the nominal diameter of the spherical ground joint.
- Not self-supporting
- Glass articles for further processing
Tekniske data
Tekniske data
| Quantità | 1 Ea. |
| Descrizione articolo | For the production of grease-free, flexible connections (alignment in 360° possible); Installation in glass apparatus and devices |
| Joint Size | 48 mm |
| Spessore (sistema metrico) parete | 3.2 mm |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion