Learn More
DWK Life Sciences DURAN™ Standard Cone, diamond-ground
Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Joint Size:
13 mm
17 mm
22 mm
26 mm
Dimensione della confezione:
pezzi 50
Descrizione
The conical ground joint of sleeve and core is a rigid, self-supporting joint. DURAN™ DG cores are not ground according to DIN 12249 and represent the cost-effective alternative to DURAN™ cores. Ground joints with ground grease or PTFE sleeves should always be used. KECK™ Conical ground joint clamps are the perfect supplement for securing the conical ground joint.
- Diamond cut (coarser cut quality, more roughness depth)
- Cost-effective alternative to DURAN™ cores (not according to DIN)
- Tight closure due to standard ground joint
- Glass articles for further processing
Specifica
Specifica
| Quantità | 50/Pk. |
| Descrizione articolo | Installation in glass apparatus and devices |
| Joint Size | 26 mm |
| Spessore (sistema metrico) parete | 2.0 mm |