Learn More
DWK Life Sciences DURAN™ Standard Cone, diamond-ground
Codice prodotto. p-7189258 Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Joint Size:
13 mm
17 mm
22 mm
26 mm
Dimensione della confezione:
pezzi 50
Codice prodotto. 15884558
Marca: DWK Life Sciences 286154208
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Designed for a rigid, self-supporting connection with lower requirements.
The conical ground joint of sleeve and core is a rigid, self-supporting joint. DURAN™ DG cores are not ground according to DIN 12249 and represent the cost-effective alternative to DURAN™ cores. Ground joints with ground grease or PTFE sleeves should always be used. KECK™ Conical ground joint clamps are the perfect supplement for securing the conical ground joint.
  • Diamond cut (coarser cut quality, more roughness depth)
  • Cost-effective alternative to DURAN™ cores (not according to DIN)
  • Tight closure due to standard ground joint
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifica
Quantità 50/Pk.
Descrizione articolo Installation in glass apparatus and devices
Joint Size 13 mm
Spessore (sistema metrico) parete 1.5 mm