Learn More
DWK Life Sciences DURAN™ Standard Cone, finished
Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Joint Size:
4 mm
6 mm
8 mm
11 mm
13 mm
17 mm
22 mm
26 mm
30 mm
36 mm
40 mm
46 mm
50 mm
56 mm
65 mm
80 mm
90 mm
Dimensione della confezione:
Pezzo
pezzi 10
pezzi 50
Descrizione
The conical ground joint is the most common joint in glass apparatus engineering. Ground DURAN™ cores comply with DIN 12249 Form B in their design and meet higher requirements for surface roughness. This reduces the need for grinding grease and the risk of jamming. Ground joints with ground grease or PTFE sleeves should always be used. KECK™ Conical ground joint clamps are the perfect supplement for securing the conical ground joint.
- According to DIN 12249 Form B
- Ground (finer grinding quality, less roughness depth)
- The finer surface reduces the risk of jamming the cuts.
- Minimized grease requirement
- Tight closure due to standard ground joint
- Glass articles for further processing
Specifica
Specifica
| Quantità | 10/Pk. |
| Descrizione articolo | Installation in glass apparatus and devices |
| Joint Size | 56 mm |
| Spessore (sistema metrico) parete | 2.5 mm |