Learn More
DWK Life Sciences DURAN™ Standard Cone with tip, finished
Codice prodotto. 15834598 Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Joint Size:
6 mm
8 mm
11 mm
13 mm
17 mm
22 mm
26 mm
30 mm
36 mm
40 mm
46 mm
50 mm
56 mm
65 mm
80 mm
90 mm
Dimensione della confezione:
Pezzo
pezzi 10
pezzi 50
Codice prodotto. 15834598
Marca: DWK Life Sciences 286171607
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Core cut with extension.
The DURAN™ cores with outlet comply with DIN 12249 Form G. The process has the function of a drip ring. It prevents the liquid from running down the inner wall of the vessel. For this purpose, the outlet is usually cut at an angle and fused. At the same time, mediums do not come into contact with the greased ground surface. The additional outlet can also be used as an attachment point for extensions for various purposes, e.g. as an inlet pipe.
  • Ground (finer grinding quality, less roughness depth)
  • The additional drain on the grinding enables a clean drain/leakage of liquids without wetting the grinding.
  • Tight closure due to standard ground joint
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifica
Quantità 10/Pk.
Descrizione articolo The additional outlet can also be used as an attachment point for extensions for various purposes, e.g. as an inlet pipe; Installation in glass apparatus and devices
Joint Size 65 mm
Spessore (sistema metrico) parete 3.2 mm