Learn More
DWK Life Sciences DURAN™ Standard Socket, diamond-ground
Codice prodotto. 15824818 Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Joint Size:
13 mm
15 mm
17 mm
22 mm
28 mm
33 mm
Dimensione della confezione:
pezzi 50
Codice prodotto. 15824818
Marca: DWK Life Sciences 286541202
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
For a rigid, self-supporting connection as a cost-effective alternative.
The conical ground joint of sleeve and core is a rigid, self-supporting joint. The DURAN™ DG sleeves are not ground according to DIN 12249 and represent a cost-effective alternative to the DURAN™ sleeves. In principle, ground joints should be made using ground grease or PTFE sleeves. KECK™ Conical ground joint clamps are the perfect supplement for securing the conical ground joint.
  • Diamond cut (coarser cut quality, more roughness depth)
  • Cost-effective alternative to DURAN™ sleeves (not according to DIN)
  • Tight closure due to standard ground joint
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifica
Quantità 50/Pk.
Descrizione articolo Use in the high vacuum range; Installation in glass apparatus and devices
Joint Size 22 mm
Spessore (sistema metrico) parete 1.8 mm