Learn More
DWK Life Sciences DURAN™ Standard Socket, diamond-ground
Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Joint Size:
13 mm
15 mm
17 mm
22 mm
28 mm
33 mm
Dimensione della confezione:
pezzi 50
Description
The conical ground joint of sleeve and core is a rigid, self-supporting joint. The DURAN™ DG sleeves are not ground according to DIN 12249 and represent a cost-effective alternative to the DURAN™ sleeves. In principle, ground joints should be made using ground grease or PTFE sleeves. KECK™ Conical ground joint clamps are the perfect supplement for securing the conical ground joint.
- Diamond cut (coarser cut quality, more roughness depth)
- Cost-effective alternative to DURAN™ sleeves (not according to DIN)
- Tight closure due to standard ground joint
- Glass articles for further processing
Specifications
Specifications
| Quantità | 50/Pk. |
| Descrizione articolo | Use in the high vacuum range; Installation in glass apparatus and devices |
| Joint Size | 17 mm |
| Spessore (sistema metrico) parete | 1.8 mm |