Learn More
DWK Life Sciences DURAN™ Standard Socket, double, diamond-ground
Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Joint Size:
18 mm
28 mm
34 mm
Dimensione della confezione:
Pezzo
pezzi 50
Descrizione
DURAN™ DG double sleeves are not ground according to DIN 12249 and represent the cost-effective alternative to DURAN™ cores.
- Diamond cut (coarser cut quality, more roughness depth)
- Cost-effective alternative to DURAN™ sleeves (not according to DIN)
- Glass article with two sleeves enables material savings
- Tight closure due to standard ground joint
- Glass articles for further processing
Specifica
Specifica
| Quantità | 25/Pk. |
| Descrizione articolo | Installation in glass apparatus and devices |
| Joint Size | 34 mm |
| Spessore (sistema metrico) parete | 2 mm |