Learn More
DWK Life Sciences DURAN™ Standard Socket, double, finished
Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Joint Size:
18 mm
22 mm
Dimensione della confezione:
pezzi 50
Descrizione
The DURAN™ double cores, ground, are an old, less common type of design according to DIN 12249 Form C. It primarily serves the purpose of saving material.
- According to DIN 12249 Form C
- Glass article with two sleeves enables material savings
- Very low leakage rate
- Minimized grease requirement
- Tight closure due to standard ground joint
- Glass articles for further processing
Specifica
Specifica
| Quantità | 50/Pk. |
| Descrizione articolo | Installation in glass apparatus and devices |
| Joint Size | 22 mm |
| Spessore (sistema metrico) parete | 1.8 mm |