Learn More
DWK Life Sciences DURAN™ Tube with Screw Thread, with DIN thread, and standard ground cone
Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Nessun giunto rastremato standard:
GL 14, with Cone NS 14/23
GL 14, with Cone NS 19/26
GL 14, with Cone NS 24/29
GL 14, with Cone NS 29/32
GL 18, with Cone NS 14/23
GL 18, with Cone NS 19/26
GL 18, with Cone NS 24/29
GL 18, with Cone NS 29/32
GL 25, with Cone NS 19/26
GL 25, with Cone NS 24/29
GL 25, with Cone NS 29/32
GL 32, with Cone NS 24/29
GL 32, with Cone NS 29/32
Dimensione della confezione:
Pezzo
pezzi 10
Descrizione
This threaded tube has a screw thread and a standard ground joint. Components of an apparatus can be connected via both ends and thus connected to each other.
- Tight closure due to standard ground joint
- With DIN thread
Specifica
Specifica
| Quantità | 10/Pk. |
| Descrizione articolo | Tube with screw threads |
| Tipo | Non-DIN Size |
| Nessun giunto rastremato standard | GL 14, with Cone NS 29/32 |