Learn More
DWK Life Sciences DURAN™ Twin-Neck Round Bottom Flask, with standard ground joint, angled side neck
Codice prodotto. p-7189463 Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Capacità (sistema metrico):
100 mL
250 mL
500 mL
1000 mL
2000 mL
Giunto rastremato standard:
NS 24/29
NS 29/32
Dimensione della confezione:
Pezzo
Codice prodotto. 10777481
Marca: DWK Life Sciences 241832707
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Multi-neck pellets are used for distillation, fraction and reactions of all kinds.
For distillation, fraction and reaction, mainly multi-neck flasks (two or three neck flasks) are used to be able to install attachments, columns, thermometers, dropping funnels, boiling capillaries, etc. as required.
  • Tight closure due to standard ground joint
  • angled side necks
  • Depending upon the application, accessories, columns, thermometers, dropping funnels, boiling capillaries, etc. can be fitted
  • The geometry permits very uniform heating
  • Fired-on, highly durable white print
TRUSTED_SUSTAINABILITY
Specifica
Diametro (sistema metrico) base 64 mm
Capacità (sistema metrico) 100 mL
Materiale Borosilicate Glass (3.3)
Da utilizzare con (applicazione) Distillation; Extraction; Connection to other glass vessels; Heating; Suitable for heating baths and mantles; Volumen sizes 100 mL (NS 24/29 and NS 29/32), 250 mL (NS 24/29), 500 mL (NS 24/29) and 1000 mL (NS 24/29) are not according to DIN ISO
N. per confezione 1/Pk.
Product Type Non-DIN/ISO size
Giunto a bussola 29/32
Giunto rastremato standard NS 29/32