Learn More
DWK Life Sciences DURAN™ Two Way Stopcock, double oblique bore, ground, without key
Codice prodotto. 15854728 Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Foro spina:
2.5 mm
4 mm
Dimensione della confezione:
pezzi 10
Codice prodotto. 15854728
Marca: DWK Life Sciences 286423006
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Ground 2-way valves without plug.
Ground DURAN™ patent valves are 2-way valves, which are frequently used for the construction of steering lines (vacuum inert gas lines, nitrogen distribution rakes). We recommend the use of PRODURAN™ valves for particularly high leakage rate requirements.
  • Valve sleeve ground, without plug
  • 2-way valve
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifica
Foro spina 2.5 mm
Diametro (sistema metrico) 8 mm
Dimensione chiusura 14.5 NS
Larghezza (sistema metrico) 1.5 mm
Da utilizzare con (applicazione) Suitable for installation in glass apparatus for vacuum applications, e.g. for the construction of steering lines (vacuum inert gas lines, nitrogen distribution rakes)