Learn More
DWK Life Sciences™ DURAN™ Vigreux Column, with 2 standard ground joints, complete, with slide-on glass jacket
Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Giunto rastremato standard:
NS 24/29
NS 29/32
Lunghezza (sistema metrico):
450 mm
650 mm
Dimensione della confezione:
Pezzo
Descrizione
Vigreux columns are used in fractionated distillation for improved separation of substances. The slightly downward angled indentations considerably increase the contact surface between the gas phase and the glass, i.e. the cooling surface.
- Tight closure due to standard ground joint
- With slide-on glass jacket
Specifica
Specifica
| Da utilizzare con (applicazione) | Fractional distillation; Laboratory coolers, condensation of gases and vapors |
| Lunghezza (sistema metrico) | 650 mm |
| Materiale | Boro 3.3 |
| Quantità | 1 Ea. |
| Giunto rastremato standard | NS 24/29 |
| Autoclavabile | Autoclavable |