Learn More
3M™ E-A-R™ E-A-Rsoft™ Yellow Neons Probed Test Plugs
Click to view available options
:
Color: Yellow
Descrizione
- These test earplugs are modified with a probe tube for use during earplug fit testing with the 3M™ E-A-Rfit™ Validation System
- Test plugs can not be used for hearing protection
- Probed test earplug for use with 3M™ E-A-Rfit™ Validation System
Specifica
Specifica
| Colore | Yellow |
| Size | Universal |
| Tipo di imballaggio | Polybag |
| Pairs per Package | 50 Pairs/Box |
| Noise Reduction Rating (NRR) | 33 dB |
| Plug Shape | Tapered |
| Product Type | Ear Plugs |
| Dielectric | No |
| Materiale | Polyurethane |