Learn More
Abeba™ Easy 37622 Shoes
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
Dimensione della confezione:
1 paio
Descrizione
- CE EN ISO 20347:2012 OB A, E, FO SRC
- Smooth leather white/blue
- Slip resistance TPU outsole sole
- Replaceable acc Wave insole (article no. 3573)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Heel strap non-foldable, adjustable
Specifica
Specifica
| Certificazione/Conformità | CE EN ISO 20347:2012 OB A, E, FO SRC |
| Colore | White |
| Materiale | Leather |
| Size | 35 |
| Caratteristiche suola | Slip-resistant |
| Materiale suola | Polyurethane |
| Stile | 37622 |