Learn More
Greiner Bio-One EasyLoad™ 1000 Refill Universal Pipette Tips
Click to view available options
:
Volume: 100 to 1000 μL
Descrizione
- Autoclavable EasyLoad™ racks
- Racks are stackable
- Color-coded boxes
- Design of rack's interior increases stability during autoclaving and when pipette tips are being used with multi-channel pipettes
Specifica
Specifica
| Volume (sistema metrico) | 100 to 1000 μL |
| Sterilità | Non sterile |
| Colore | Blue |
| Autoclavabile | Autoclavable |
| Formato | Ricarica |
| Da utilizzare con (apparecchiatura) | Eppendorf, Thermo Scientific Finnpipette, Gilson, Socorex |
| Graduato | Yes |
| Product Line | EasyLoad |
| Materiale | Polipropilene |