Learn More
Terumo™ Siringa concentrica Luer-Lock in tre parti
Terumo™ Eccentric Luer-Lock Three-Part Syringes are easy to use with their smooth plunger action and have clear barrels to allow quick recognition of air bubbles for more accurate dosing. With easy to read markings, you can be confident you are delivering the intended dose every time.
Marca: Terumo™ 1SS02LE1
Detalles adicionales : Peso : 0.35100kg
Descripción
- Silicone coated barrel for a smooth plunger action
- Clear barrel to allow quick recognition of air bubbles
- Easy to read markings
- Latex free
- Sterile and packed in individually wrapped tamper peel packs
Especificaciones
Sì | |
Graduazione serigrafata | |
Sì | |
Sì | |
2.5 ml | |
Sì |
Polypropylene | |
Without needle | |
Siringa attacco centrico Luer Lock a 3 parti | |
Confezione da 100 | |
Luer Lock centrico |