Learn More
Bellingham + Stanley™ Rifrattometro Brix manuale ottico di livello professionale
GREENER_CHOICE
Codice prodotto. 12303539
Click to view available options
Scale Range:
0 - 50% Brix
0 to 15 Brix
0 to 30 Brix
0 to 50 Brix
28 to 65 Brix
45 - 80% Brix
45 to 80 Brix
72 to 95 Brix
Incrementi:
0.1
0.2
0.5
Tipo di bilancia:
Brix
Low Volume Brix
Refractive Index
Dimensione della confezione:
Pezzo
Codice prodotto. 12303539
Marca: Bellingham + Stanley™ 4582
Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Visualizzare prodotti alternativi
Manufactured to the highest specification using only the best materials and optical components. Bellingham + Stanley™ Eclipse Hand Held Refractometer is a low-cost, portable instrument used for on-the-spot determination of Refractive Index of liquids or the concentration of solutions.
The Eclipse professional-grade range of scales are available for general use in a variety of applications, with special models available, such as Low Volume Brix models to facilitate measurement of samples of less than 1μL that are used by botanists and zoologists for measuring nectar. All instruments are supplied with a soft carrying case and instructions for use.
These reliable and robust refractometers are sealed against the ingress of water with IP65 construction, so they can be used in the harshest of environments. The large body housing and rubber handgrip make the instrument incredible easy to handle. Special features make measurements easy and reliable:
  • Simple zero adjust with lock
  • Shaded view with bright display for maximum distinction
  • High precision, clear scale
  • Easy-clean prism
  • Unique sample ‘dribble feed’ feature
  • Low weight
  • Anti-roll supports
  • Push-on prism flap
TRUSTED_SUSTAINABILITY
Specifica
Tipo Eclipse Professional Grade Optical Hand Held Refractometer
Scale Range 45 to 80 Brix
Volume del campione <1 μL
Incrementi 0.2
Tipo di bilancia Low Volume Brix
Da utilizzare con (applicazione) ‘On the spot’ determination of Refractive Index (RI) of liquids or the concentration of solutions
Include Soft carrying case
Protezione IP IP65