Learn More
Schuett Biotec™ EcoVac Safety Extraction System 4
Codice prodotto. 10308961
Click to view available options
Portata:
4 L/min.
Dimensione della confezione:
Pezzo
Codice prodotto. 10308961
Marca: Schuett Biotec™ 3350042
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
The EcoVac Safety Extraction System 4 is for safe and convenient aspiration of liquid supernatants in cell culture. Increases work safety when handling infectious material. Autoclavable, unbreakable and resistant to chemicals.
  • For aspiration of liquid supernatants in cell culture
  • Increases work safety
  • Completely autoclavable, unbreakable and resistant to chemicals
  • Quick-release couplings automatically close the bottle and hermetically seal it for transport
Specifica
Vuoto max 300 mbar
Include EcoVac system 4, 4 L safety collection bottle with screw cap, 2 quick-release couplings, 2 angle nipples for hose connections, 2 x 1.5 m silicone hose, ventilation insert and safety filter
Portata 4 L/min.