missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Efficient-Pro™ AGT™ Medium
Click to view available options
Quantità:
1 L
10 L
100 L
500 L
Dimensione della confezione:
100L
10L
1L
500L
Descrizione
- A next generation CHO media used in the production of monoclonal antibodies
- AGT Medium offers significant performance and ease-of-use improvements not seen in existing Gibco™ or competitor products
- Proprietary AGT™ format is the only one of its kind in the industry and is the most advanced media format available in the market
- Formulated as a chemically-defined, animal-origin free cell culture media used in the bioproduction of mAbs & other therapeutics in traditional batch & fed-batch bioprocessing workflows
- Free of growth factors and insulin to ensure batch-to-batch consistency and production efficiency
- Designed to work optimally with Efficient-Pro Feed 1 and Efficient-Pro Feed 2 for use in CHO-K1 and DG-44/CHO-S cell lines respectively
- The Efficient-Pro system increases titer and specific productivity, improves protein quality and metabolite profiles
- System is easier to use with less solubility issues than other CHO media and feed offerings
- Increase ease-of-use with Gibco’s™ trademarked, patented Advanced Granulation Technology™ (AGT™) for auto-pH and simplified dissolution
- Sustain cell viability for longer bioreactor runs & maximized titer
- Demonstrated to improve titers in a CHO-K1 cell line up to 147% when paired with Efficient-Pro™ Feed 1 compared to an industry-leader’s media and feed system
- Improve downstream outputs such as protein glycosylation, charge-variants & aggregation
- Speed-up process development and streamline transfer to manufacturing scale
- Increase flexibility with a range of format & packaging configurations
- Decrease cell shock with simplified adaptation protocols
Specifica
Specifica
| Linea cellulare | CHO-DG44, CHO-S, CHO-K1, GS-CHO |
| Classificazione | Animal Origin-free, Chemically-defined, Protein-free, Serum-free |
| Forma | Powder (AGT) |
| Quantità | 10 L |
| Shipping Condition | Room temperature |
| Green Features | Sustainable packaging |
| Content And Storage | Store in the dark at 2–8°C. |
| Culture Type | Mammalian Cell Culture |
For research use only. Not for use in diagnostic procedures.
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto